siRNA Details
| Gene Symbol: Avl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030235.1 | NP_084511.1 | 16 | 3172 | ||
| GenBank mRNA | AK083605.1 | - | 16 | 3129 | ||
| AK122228.1 | - | 16 | 3150 | |||
| AK147365.1 | - | 16 | 3173 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Avl9| Gene Symbol: Avl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030235.1 | NP_084511.1 | 16 | 3172 | ||
| GenBank mRNA | AK083605.1 | - | 16 | 3129 | ||
| AK122228.1 | - | 16 | 3150 | |||
| AK147365.1 | - | 16 | 3173 | |||