siRNA Details
| Gene Symbol: Tomt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081679.1 | NP_001075148.1 | 4 | 1007 | ||
| NM_001282088.1 | NP_001269017.1 | 4 | 1016 | |||
| GenBank mRNA | AK158024.1 | - | 1017 | |||
| DQ854743.1 | - | 4 | 1007 | |||
| EU627091.1 | - | 4 | 1016 | |||

Tomt
Genomic Map