siRNA Details
| Gene Symbol: Slc25a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159275.1 | NP_001152747.1 | 1 | 1204 | ||
| GenBank mRNA | AF332006.1 | - | 1 | 1062 | ||
| AK077159.1 | - | 1 | 1205 | |||
| AY583456.1 | - | 1 | 1189 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a2| Gene Symbol: Slc25a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159275.1 | NP_001152747.1 | 1 | 1204 | ||
| GenBank mRNA | AF332006.1 | - | 1 | 1062 | ||
| AK077159.1 | - | 1 | 1205 | |||
| AY583456.1 | - | 1 | 1189 | |||