siRNA Details
| Gene Symbol: C87977 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177542.1 | NP_001171013.1 | 4 | 1571 | ||
| GenBank mRNA | AK139502.1 | - | 4 | 1570 | ||
| BC094895.1 | - | 4 | 1542 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C87977| Gene Symbol: C87977 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177542.1 | NP_001171013.1 | 4 | 1571 | ||
| GenBank mRNA | AK139502.1 | - | 4 | 1570 | ||
| BC094895.1 | - | 4 | 1542 | |||