siRNA Details
Gene Symbol: C86695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081662.1 | NP_001075131.1 | 5 | 872 | ||
GenBank mRNA | AK135805.1 | - | 5 | 871 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C86695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081662.1 | NP_001075131.1 | 5 | 872 | ||
GenBank mRNA | AK135805.1 | - | 5 | 871 |