siRNA Details
| Gene Symbol: Khdc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113187.1 | NP_001106658.1 | 1 | 209 | ||
| GenBank mRNA | AK139842.1 | - | 1 | 209 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Khdc1b| Gene Symbol: Khdc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113187.1 | NP_001106658.1 | 1 | 209 | ||
| GenBank mRNA | AK139842.1 | - | 1 | 209 | ||