siRNA Details
Gene Symbol: Syt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019350.2 | NP_062223.1 | 3, 4 | 581 | ||
XM_006228287.2 | XP_006228349.1 | 2, 3 | 271 | |||
GenBank mRNA | BC092198.1 | - | 3, 4 | 581 | ||
U26402.1 | - | 3, 4 | 384 | |||
X84884.1 | - | 2, 3 | 334 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
