siRNA Details
| Gene Symbol: Qser1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123327.2 | NP_001116799.1 | 12 | 6185 | ||
| GenBank mRNA | BC021511.1 | - | 1 | 596 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Qser1| Gene Symbol: Qser1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123327.2 | NP_001116799.1 | 12 | 6185 | ||
| GenBank mRNA | BC021511.1 | - | 1 | 596 | ||