siRNA Details
| Gene Symbol: Cep152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081091.1 | NP_001074560.1 | 6 | 805 | ||
| GenBank mRNA | AK030816.1 | - | 1 | 52 | ||
| AK041913.1 | - | 6 | 794 | |||
| AK173060.1 | - | 6 | 805 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep152| Gene Symbol: Cep152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081091.1 | NP_001074560.1 | 6 | 805 | ||
| GenBank mRNA | AK030816.1 | - | 1 | 52 | ||
| AK041913.1 | - | 6 | 794 | |||
| AK173060.1 | - | 6 | 805 | |||