siRNA Details
| Gene Symbol: AU015228 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033197.2 | NP_001028369.1 | 1 | 1865 | ||
| GenBank mRNA | AK139778.1 | - | 1 | 1866 | ||
| AK163392.1 | - | 1 | 1867 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AU015228| Gene Symbol: AU015228 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033197.2 | NP_001028369.1 | 1 | 1865 | ||
| GenBank mRNA | AK139778.1 | - | 1 | 1866 | ||
| AK163392.1 | - | 1 | 1867 | |||