siRNA Details
| Gene Symbol: Clca4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033199.3 | NP_001028371.1 | 14 | 2471 | ||
| GenBank mRNA | AK136515.1 | - | 14 | 2476 | ||
| BC139088.1 | - | 14 | 2465 | 8862110 | ||
| BC139089.1 | - | 14 | 2465 | 8862111 | ||
| JN652132.1 | - | 14 | 2442 | |||

Clca4b
Genomic Map