siRNA Details
| Gene Symbol: Tpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019353.2 | NP_062226.2 | 5 | 426 | ||
| XM_008764607.1 | XP_008762829.1 | 4 | 384 | |||
| XM_008764608.1 | XP_008762830.1 | 5 | 420 | |||
| GenBank mRNA | X17396.1 | - | 5 | 425 | ||

Tpo
Genomic Map