siRNA Details
| Gene Symbol: Stk39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019362.1 | NP_062235.1 | 3, 4 | 881 | ||
| XM_008761922.1 | XP_008760144.1 | 3, 4 | 625 | |||
| GenBank mRNA | AF068261.1 | - | 3, 4 | 646 | ||
| AF099990.1 | - | 3, 4 | 585 | |||
| D88190.1 | - | 3, 4 | 881 | |||

Stk39
Genomic Map