siRNA Details
| Gene Symbol: Ywhab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019377.1 | NP_062250.1 | 2 | 256 | ||
| GenBank mRNA | BC076502.1 | - | 2 | 250 | 7100393 | |
| D17446.1 | - | 2 | 251 | |||
| FQ214109.1 | - | 2 | 292 | |||
| FQ229978.1 | - | 2 | 261 | |||
| S55223.1 | - | 2 | 251 | |||
| S83440.1 | - | 2 | 256 | |||

Ywhab
Genomic Map