siRNA Details
| Gene Symbol: Gip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019630.3 | NP_062604.1 | 2, 3 | 129 | ||
| XM_006247177.2 | XP_006247239.1 | 1, 2 | 351 | |||
| XM_008767947.1 | XP_008766169.1 | 2, 3 | 171 | |||
| GenBank mRNA | D11439.1 | - | 1, 2 | 101 | ||
| L08831.1 | - | 2, 3 | 129 | |||
| X66724.1 | - | 2, 3 | 190 | |||
| Z19564.1 | - | 2, 3 | 129 | |||

Gip
Genomic Map