siRNA Details
| Gene Symbol: Mtor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019906.1 | NP_063971.1 | 2 | 197 | ||
| GenBank mRNA | L37085.1 | - | 1 | 161 | ||
| U11681.1 | - | 2 | 197 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtor| Gene Symbol: Mtor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019906.1 | NP_063971.1 | 2 | 197 | ||
| GenBank mRNA | L37085.1 | - | 1 | 161 | ||
| U11681.1 | - | 2 | 197 | |||