siRNA Details
| Gene Symbol: Eif5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020075.1 | NP_064460.1 | 1, 2 | 307 | ||
| XM_006240608.1 | XP_006240670.1 | 3, 4 | 570 | |||
| XM_006240609.2 | XP_006240671.1 | 2, 3 | 850 | |||
| GenBank mRNA | BC062398.1 | - | 2, 3 | 495 | 5623373 | |
| L11651.1 | - | 1, 2 | 307 | |||

Eif5
Genomic Map