siRNA Details
| Gene Symbol: Rnase4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020082.2 | NP_064467.1 | 2 | 255 | ||
| XM_006251906.2 | XP_006251968.1 | 2 | 294 | |||
| XM_006251907.2 | XP_006251969.1 | 2 | 3872 | |||
| GenBank mRNA | AF041066.1 | - | 2 | 266 | ||
| BC070953.1 | - | 2 | 255 | 7103267 | ||
| BC166436.1 | - | 2 | 838 | 7372999 | ||

Rnase4
Genomic Map