siRNA Details
| Gene Symbol: Timm23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019352.2 | NP_062225.1 | 3 | 320 | ||
| GenBank mRNA | AB006451.1 | - | 3 | 320 | ||
| BC083559.1 | - | 3 | 327 | 7135081 | ||
| FQ216304.1 | - | 3 | 324 | |||
| FQ229692.1 | - | 3 | 324 | |||
| FQ229803.1 | - | 3 | 326 | |||

Timm23
Genomic Map