siRNA Details
| Gene Symbol: Bet1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019368.3 | NP_062241.2 | 2, 3 | 157 | ||
| XM_006230574.2 | XP_006230636.1 | 3, 4 | 369 | |||
| GenBank mRNA | AF003998.1 | - | 2, 3 | 196 | ||
| BC059138.1 | - | 2, 3 | 161 | 6890305 | ||
| FQ228853.1 | - | 3, 4 | 319 | |||

Bet1l
Genomic Map