siRNA Details
| Gene Symbol: Acpp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134901.1 | NP_001128373.1 | 2 | 182 | ||
| NM_020072.1 | NP_064457.1 | 2 | 222 | |||
| GenBank mRNA | DQ826426.1 | - | 2 | 182 | ||
| M32397.1 | - | 2 | 222 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acpp| Gene Symbol: Acpp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134901.1 | NP_001128373.1 | 2 | 182 | ||
| NM_020072.1 | NP_064457.1 | 2 | 222 | |||
| GenBank mRNA | DQ826426.1 | - | 2 | 182 | ||
| M32397.1 | - | 2 | 222 | |||