siRNA Details
| Gene Symbol: Nr4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019328.3 | NP_062201.2 | 5 | 1424 | ||
| XM_006234201.2 | XP_006234263.1 | 6 | 1642 | |||
| XM_008761861.1 | XP_008760083.1 | 5 | 1558 | |||
| GenBank mRNA | DQ092629.1 | - | 5 | 1355 | ||
| DQ092630.1 | - | 5 | 1191 | |||
| DQ092631.1 | - | 5 | 1369 | |||
| DQ092632.1 | - | 5 | 1359 | |||
| DQ092633.1 | - | 5 | 1191 | |||
| DQ092634.1 | - | 5 | 1191 | |||
| FQ222109.1 | - | 6 | 1664 | |||
| L08595.1 | - | 5 | 1421 | |||
| U01146.1 | - | 5 | 1355 | |||
| U72345.1 | - | 6 | 1648 | |||

Nr4a2
Genomic Map