siRNA Details
| Gene Symbol: Rgs5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019341.1 | NP_062214.1 | 4, 5 | 392 | ||
| GenBank mRNA | AF241259.1 | - | 4, 5 | 392 | ||
| EF568934.1 | - | 4, 5 | 456 | |||
| FQ234716.1 | - | 4, 5 | 509 | |||
| U32435.1 | - | 1, 2 | 110 | |||

Rgs5
Genomic Map