siRNA Details
| Gene Symbol: Rgs7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019343.1 | NP_062216.1 | 14 | 1222 | ||
| GenBank mRNA | AB024398.1 | - | 14 | 1222 | ||
| U32328.1 | - | 1 | 65 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rgs7| Gene Symbol: Rgs7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019343.1 | NP_062216.1 | 14 | 1222 | ||
| GenBank mRNA | AB024398.1 | - | 14 | 1222 | ||
| U32328.1 | - | 1 | 65 | |||