siRNA Details
| Gene Symbol: Slc14a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019346.2 | NP_062219.2 | 5 | 690 | ||
| XM_008772168.1 | XP_008770390.1 | 6 | 908 | |||
| XM_008772169.1 | XP_008770391.1 | 5 | 1822 | |||
| XM_008772170.1 | XP_008770392.1 | 5 | 759 | |||
| GenBank mRNA | FQ211495.1 | - | 5 | 723 | ||
| U81518.1 | - | 5 | 660 | |||
| X98399.1 | - | 5 | 690 | |||

Slc14a1
Genomic Map