siRNA Details
| Gene Symbol: Tpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019353.2 | NP_062226.2 | 12 | 2066 | ||
| XM_008764607.1 | XP_008762829.1 | 11 | 2024 | |||
| XM_008764608.1 | XP_008762830.1 | 12 | 2060 | |||
| GenBank mRNA | M31655.1 | - | 8 | 1592 | ||
| X17396.1 | - | 12 | 2065 | |||

Tpo
Genomic Map