siRNA Details
| Gene Symbol: Pdpn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019358.1 | NP_062231.1 | 2 | 413 | ||
| XM_006239338.2 | XP_006239400.1 | 2 | 413 | |||
| GenBank mRNA | BC072492.1 | - | 2 | 369 | 7097186 | |
| U07797.1 | - | 2 | 413 | |||
| U32115.1 | - | 2 | 341 | |||
| U96449.1 | - | 2 | 193 | |||

Pdpn
Genomic Map