siRNA Details
| Gene Symbol: Anxa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019905.1 | NP_063970.1 | 6, 7 | 494 | ||
| GenBank mRNA | BC059136.1 | - | 6, 7 | 513 | 6920034 | |
| FQ219918.1 | - | 1, 2 | 60 | |||
| FQ225322.1 | - | 6, 7 | 510 | |||
| FQ225353.1 | - | 3, 4 | 282 | |||
| L13039.1 | - | 5, 6 | 465 | |||
| S73559.1 | - | 6, 7 | 496 | |||
| X66871.1 | - | 6, 7 | 494 | |||

Anxa2
Genomic Map