siRNA Details
| Gene Symbol: Srgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020074.2 | NP_064459.1 | 3 | 261 | ||
| XM_006256483.2 | XP_006256545.1 | 6 | 641 | |||
| XM_006256484.2 | XP_006256546.1 | 4 | 449 | |||
| GenBank mRNA | BC088144.1 | - | 3 | 261 | 7384101 | |
| FQ220127.1 | - | 3 | 280 | |||
| FQ227535.1 | - | 3 | 278 | |||
| FQ231972.1 | - | 3 | 278 | |||
| J03224.1 | - | 3 | 274 | |||
| K02934.1 | - | 3 | 276 | |||

Srgn
Genomic Map