siRNA Details
| Gene Symbol: Haao | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020076.2 | NP_064461.1 | 5 | 468 | ||
| XM_006239684.2 | XP_006239746.1 | 5 | 1373 | |||
| GenBank mRNA | BC085739.1 | - | 5 | 422 | 7108551 | |
| D28339.1 | - | 3 | 182 | |||
| D44494.1 | - | 5 | 469 | |||
| FQ219702.1 | - | 5 | 482 | |||

Haao
Genomic Map