siRNA Details
| Gene Symbol: Cd86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020081.1 | NP_064466.1 | 4 | 452 | ||
| XM_006248399.2 | XP_006248461.1 | 4 | 819 | |||
| XM_006248400.1 | XP_006248462.1 | 3 | 428 | |||
| GenBank mRNA | D50558.1 | - | 4 | 452 | ||
| U31330.1 | - | 2 | 298 | |||

Cd86
Genomic Map