siRNA Details
| Gene Symbol: Ifit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020096.1 | NP_064481.1 | 1 | 1279 | ||
| GenBank mRNA | AJ276893.1 | - | 1 | 1279 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifit1| Gene Symbol: Ifit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020096.1 | NP_064481.1 | 1 | 1279 | ||
| GenBank mRNA | AJ276893.1 | - | 1 | 1279 | ||