siRNA Details
| Gene Symbol: Adap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020101.1 | NP_064486.1 | 6 | 582 | ||
| GenBank mRNA | AJ238993.1 | - | 6 | 582 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adap2| Gene Symbol: Adap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020101.1 | NP_064486.1 | 6 | 582 | ||
| GenBank mRNA | AJ238993.1 | - | 6 | 582 | ||