siRNA Details
| Gene Symbol: Adam17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020306.2 | NP_064702.1 | 2 | 414 | ||
| GenBank mRNA | AJ012603.1 | - | 2 | 416 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adam17| Gene Symbol: Adam17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020306.2 | NP_064702.1 | 2 | 414 | ||
| GenBank mRNA | AJ012603.1 | - | 2 | 416 | ||