siRNA Details
| Gene Symbol: Acp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021262.2 | NP_067085.1 | 3 | 223 | ||
| XM_006239965.2 | XP_006240027.1 | 3 | 252 | |||
| GenBank mRNA | AF171072.1 | - | 3 | 186 | ||
| BC062229.1 | - | 3 | 223 | 6918194 | ||
| FQ213063.1 | - | 3 | 203 | |||
| FQ214806.1 | - | 2 | 91 | |||

Acp1
Genomic Map