siRNA Details
| Gene Symbol: Chga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021655.2 | NP_067687.2 | 2, 3 | 252 | ||
| GenBank mRNA | AF145445.1 | - | 2, 3 | 114 | ||
| BC087703.1 | - | 2, 3 | 232 | 7313842 | ||
| CB743841.1 | - | 2, 3 | 308 | |||
| X06832.1 | - | 2, 3 | 237 | |||

Chga
Genomic Map