siRNA Details
| Gene Symbol: Hcn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021658.1 | NP_067690.1 | 1 | 319 | ||
| GenBank mRNA | AF247453.1 | - | 319 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hcn4| Gene Symbol: Hcn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021658.1 | NP_067690.1 | 1 | 319 | ||
| GenBank mRNA | AF247453.1 | - | 319 | |||