siRNA Details
| Gene Symbol: F13a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021698.2 | NP_067730.2 | 3 | 245 | ||
| GenBank mRNA | DN931132.1 | - | 3 | 651 | ||
| Y12502.1 | - | 3 | 212 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

F13a1| Gene Symbol: F13a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021698.2 | NP_067730.2 | 3 | 245 | ||
| GenBank mRNA | DN931132.1 | - | 3 | 651 | ||
| Y12502.1 | - | 3 | 212 | |||