siRNA Details
| Gene Symbol: Ppp3r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021701.2 | NP_067733.1 | 1 | 91 | ||
| GenBank mRNA | D10393.1 | - | 1 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp3r2| Gene Symbol: Ppp3r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021701.2 | NP_067733.1 | 1 | 91 | ||
| GenBank mRNA | D10393.1 | - | 1 | 94 | ||