siRNA Details
| Gene Symbol: Nr5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021742.1 | NP_068510.1 | 2 | 174 | ||
| GenBank mRNA | AB012960.1 | - | 2 | 174 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nr5a2| Gene Symbol: Nr5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021742.1 | NP_068510.1 | 2 | 174 | ||
| GenBank mRNA | AB012960.1 | - | 2 | 174 | ||