siRNA Details
| Gene Symbol: Acp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135562.1 | NP_001129034.1 | 6 | 531 | ||
| NM_021262.2 | NP_067085.1 | 6 | 498 | |||
| GenBank mRNA | AF171072.1 | - | 461 | |||
| BC062229.1 | - | 6 | 498 | 6918194 | ||
| BC158763.1 | - | 6 | 531 | 7109165 | ||
| BC161990.1 | - | 6 | 477 | 9032684 | ||
| FQ213063.1 | - | 6 | 478 | |||
| FQ214806.1 | - | 5 | 366 | |||
| FQ215147.1 | - | 6 | 469 | |||

Acp1
Genomic Map