siRNA Details
| Gene Symbol: Rpl35a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021264.3 | NP_067087.1 | 193 | |||
| GenBank mRNA | BC061557.1 | - | 259 | 6917939 | ||
| X03475.1 | - | 193 | ||||
| Gene Symbol: LOC100359498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002727895.4 | XP_002727941.1 | 4 | 284 | ||
| XM_002727896.4 | XP_002727942.1 | 4 | 289 | |||
| GenBank mRNA | FQ210925.1 | - | 4 | 253 | ||

Rpl35a
Genomic Map