siRNA Details
| Gene Symbol: Nxf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021579.1 | NP_067590.1 | 2 | 232 | ||
| GenBank mRNA | AF093139.1 | - | 2 | 284 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nxf1| Gene Symbol: Nxf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021579.1 | NP_067590.1 | 2 | 232 | ||
| GenBank mRNA | AF093139.1 | - | 2 | 284 | ||