siRNA Details
Gene Symbol: Hand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021592.2 | NP_067603.1 | 1 | 532 | ||
GenBank mRNA | BC097324.1 | - | 1 | 532 | 7460879 | |
Y08140.1 | - | 1 | 331 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021592.2 | NP_067603.1 | 1 | 532 | ||
GenBank mRNA | BC097324.1 | - | 1 | 532 | 7460879 | |
Y08140.1 | - | 1 | 331 |