siRNA Details
| Gene Symbol: Dnase2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021664.1 | NP_067696.1 | 4 | 614 | ||
| GenBank mRNA | AF178974.1 | - | 4 | 614 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnase2b| Gene Symbol: Dnase2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021664.1 | NP_067696.1 | 4 | 614 | ||
| GenBank mRNA | AF178974.1 | - | 4 | 614 | ||