siRNA Details
| Gene Symbol: Erbb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021687.1 | NP_067719.1 | 6 | 670 | ||
| GenBank mRNA | AF041838.1 | - | 6 | 670 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Erbb4| Gene Symbol: Erbb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021687.1 | NP_067719.1 | 6 | 670 | ||
| GenBank mRNA | AF041838.1 | - | 6 | 670 | ||