siRNA Details
| Gene Symbol: Nr1h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021745.1 | NP_068513.1 | 4 | 724 | ||
| XM_008765260.1 | XP_008763482.1 | 3 | 1082 | |||
| XM_008765261.1 | XP_008763483.1 | 3 | 1081 | |||
| XM_008765262.1 | XP_008763484.1 | 4 | 763 | |||
| GenBank mRNA | BC083815.1 | - | 3 | 602 | 7110420 | |
| U18374.1 | - | 4 | 724 | |||

Nr1h4
Genomic Map