siRNA Details
| Gene Symbol: Cnr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020543.4 | NP_065418.3 | 1 | 954 | ||
| GenBank mRNA | AF218846.1 | - | 1 | 954 | ||
| FJ694960.1 | - | 2 | 1122 | |||
| FJ694961.1 | - | 2 | 1224 | |||
| JN420349.1 | - | 3 | 1319 | |||
| JX494784.1 | - | 2 | 1224 | |||
More Information
Set Membership: |
|

Cnr2
Genomic Map