siRNA Details
| Gene Symbol: Nt5e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021576.2 | NP_067587.2 | 4 | 941 | ||
| GenBank mRNA | BC081806.1 | - | 4 | 971 | 7134340 | |
| BC098665.1 | - | 4 | 941 | 7322840 | ||
| J05214.1 | - | 4 | 1291 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nt5e| Gene Symbol: Nt5e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021576.2 | NP_067587.2 | 4 | 941 | ||
| GenBank mRNA | BC081806.1 | - | 4 | 971 | 7134340 | |
| BC098665.1 | - | 4 | 941 | 7322840 | ||
| J05214.1 | - | 4 | 1291 | |||