siRNA Details
| Gene Symbol: Ltbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021586.2 | NP_067597.2 | 9 | 1912 | ||
| XM_006240350.2 | XP_006240412.1 | 9 | 2104 | |||
| GenBank mRNA | Y12760.1 | - | 9 | 1911 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ltbp2| Gene Symbol: Ltbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021586.2 | NP_067597.2 | 9 | 1912 | ||
| XM_006240350.2 | XP_006240412.1 | 9 | 2104 | |||
| GenBank mRNA | Y12760.1 | - | 9 | 1911 | ||